3,5-Dichloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine ≥95%
Supplier: Matrix Scientific
046703-500MG
046703-1G
046703-5G
101876-634EA
181.54
USD
101876-634
101876-636
101876-638
3,5-Dichloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine ≥95%
3,5-Dichloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine
Hazard: Irritant
Formula:
C₁₂H₆Cl₃F₃N₂O Melting Pt: 143…146 °C |
MDL Number:
MFCD11553026 CAS Number: 73265-15-3 |
Specification Test Results
Melting point | 143-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...