Methyl-2-[2-(chloromethyl)-1,3-benzoxazol-5-yl]acetate ≥95%

Supplier: Matrix Scientific

046702-500MG 046702-1G 046702-5G
101876-628EA 1109.23 USD
101876-628 101876-630 101876-632
Methyl-2-[2-(chloromethyl)-1,3-benzoxazol-5-yl]acetate ≥95%
Methyl-2-[2-(chloromethyl)-1,3-benzoxazol-5-yl]acetate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₃
MW: 239.66 g/mol
Melting Pt: 61…64 °C
MDL Number: MFCD08444008
CAS Number: 924869-02-3

Order Now

Specification Test Results

Melting point 61-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR