N-(4-Bromophenyl)-N'-(2-chloroethyl)urea ≥95%

Supplier: Matrix Scientific

046701-500MG 046701-1G 046701-5G
101876-622EA 278.46 USD
101876-622 101876-624 101876-626
N-(4-Bromophenyl)-N'-(2-chloroethyl)urea ≥95%
N-(4-Bromophenyl)-N'-(2-chloroethyl)urea

Hazard: Irritant

Formula: C₉H₁₀BrClN₂O
Melting Pt: 172…174 °C
MDL Number: MFCD08444007
CAS Number: 15145-38-7

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR