2-(Acetylamino)-3-[(4-bromophenyl)sulfanyl]propanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Acetamido)-3-[(4-bromophenyl)thio]propanoic acid
046700-500MG
046700-1G
046700-5G
101876-616EA
278.46
USD
101876-616
101876-618
101876-620
2-(Acetylamino)-3-[(4-bromophenyl)sulfanyl]propanoic acid ≥95%
2-(Acetylamino)-3-[(4-bromophenyl)sulfanyl]propanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₂BrNO₃S Melting Pt: 152…155 °C |
MDL Number:
MFCD00869819 CAS Number: 126253-78-9 |
Specification Test Results
Melting point | 152-155°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...