Ethyl-4-hydroxy-2-(2-thienyl)-1,3-thiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
046698-500MG
046698-1G
046698-5G
101876-604EA
278.46
USD
101876-604
101876-606
101876-608
Ethyl-4-hydroxy-2-(2-thienyl)-1,3-thiazole-5-carboxylate ≥95%
Ethyl-4-hydroxy-2-(2-thienyl)-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉NO₃S₂ MW: 255.32 g/mol Melting Pt: 128…130 °C |
MDL Number:
MFCD08444005 CAS Number: 924869-00-1 |
Specification Test Results
Melting point | 128-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...