2-[2-(4-Chlorophenyl)-5-methyl-1,3-thiazol-4-yl]acetic acid ≥95%

Supplier: Matrix Scientific

046695-500MG 046695-1G 046695-5G
101876-586EA 375.38 USD
101876-586 101876-588 101876-590
2-[2-(4-Chlorophenyl)-5-methyl-1,3-thiazol-4-yl]acetic acid ≥95%
2-[2-(4-Chlorophenyl)-5-methyl-1,3-thiazol-4-yl]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂S
MW: 267.74 g/mol
Melting Pt: 155…156 °C
MDL Number: MFCD00728866
CAS Number: 259655-35-1

Order Now

Specification Test Results

Melting point 155-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR