2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Chlorophenyl)-4-hydroxy-5-phenyl-1,3-thiazole
046690-500MG
046690-1G
046690-5G
101876-556EA
181.54
USD
101876-556
101876-558
101876-560
2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol ≥95%
2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol
Hazard: Irritant
Formula:
C₁₅H₁₀ClNOS MW: 287.77 g/mol Melting Pt: 252…254 °C |
MDL Number:
MFCD08444002 CAS Number: 65752-50-3 |
Specification Test Results
Melting point | 252-254°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...