5-Phenyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-5-phenyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole

046689-500MG 046689-1G 046689-5G
101876-550EA 181.54 USD
101876-550 101876-552 101876-554
5-Phenyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-ol ≥95%
5-Phenyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-ol

Hazard: Irritant

Formula: C₁₆H₁₀F₃NOS
MW: 321.32 g/mol
Melting Pt: 201…204 °C
MDL Number: MFCD08444001
CAS Number: 924868-98-4

Order Now

Specification Test Results

Melting point 201-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR