4-(Chloromethyl)-1-phenyl-1H-1,2,3-triazole ≥95%
Supplier: Matrix Scientific
046688-500MG
046688-1G
046688-5G
101876-544EA
400
USD
101876-544
101876-546
101876-548
4-(Chloromethyl)-1-phenyl-1H-1,2,3-triazole ≥95%
4-(Chloromethyl)-1-phenyl-1H-1,2,3-triazole
Hazard: Irritant
Formula:
C₉H₈ClN₃ Melting Pt: 113…115 °C |
MDL Number:
MFCD08444000 CAS Number: 924868-97-3 |
Specification Test Results
Melting point | 113-115°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...