4'-Nitro[1,1'-biphenyl]-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046685-500MG 046685-1G
101876-536EA 375.38 USD
101876-536 101876-538
4'-Nitro[1,1'-biphenyl]-2-carboxylic acid ≥95%
4'-Nitro[1,1'-biphenyl]-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉NO₄
Melting Pt: 228…230 °C
MDL Number: MFCD00445193
CAS Number: 18211-41-1

Order Now

Specification Test Results

Melting point 228-230°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR