Methyl-3-hydroxy-4-methoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-hydroxy-4-methoxybenzoate, Methyl isovanillate

046684-5G 046684-25G
101876-532EA 53.85 USD
101876-532 101876-534
Methyl-3-hydroxy-4-methoxybenzoate ≥95%
Methyl-3-hydroxy-4-methoxybenzoate

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Boiling Pt: 102°C/0.1 Torr
Melting Pt: 64…67 °C
MDL Number: MFCD01321262
CAS Number: 6702-50-7

Order Now

Specification Test Results

Melting point 64-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR