5-(1,3-Benzothiazol-2-yl)-2-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(1,3-Benzothiazol-2-yl)-2-hydroxypyridine

046683-500MG 046683-1G 046683-5G
101876-524EA 278.46 USD
101876-524 101876-526 101876-528
5-(1,3-Benzothiazol-2-yl)-2-pyridinol ≥95%
5-(1,3-Benzothiazol-2-yl)-2-pyridinol

Hazard: Irritant

Formula: C₁₂H₈N₂OS
MW: 228.27 g/mol
Melting Pt: 236…237 °C
MDL Number: MFCD08443999
CAS Number: 54628-01-2

Order Now

Specification Test Results

Melting point 236-237°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR