tert-Butyl-2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate ≥95%

Supplier: Matrix Scientific

046680-500MG 046680-1G 046680-5G
101876-514EA 181.54 USD
101876-514 101876-516 101876-518
tert-Butyl-2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate ≥95%
tert-Butyl-2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 60…61 °C
MDL Number: MFCD09972233
CAS Number: 114650-82-7

Order Now

Specification Test Results

Melting point 60-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR