Methyl-3-chloro-2-piperazino-5-(trifluoromethyl)isonicotinate ≥95%
Supplier: Matrix Scientific
046670-500MG
046670-1G
101876-490EA
1109.23
USD
101876-490
101876-492
Methyl-3-chloro-2-piperazino-5-(trifluoromethyl)isonicotinate ≥95%
Methyl-3-chloro-2-piperazino-5-(trifluoromethyl)isonicotinate
Hazard: Irritant
Formula:
C₁₂H₁₃ClF₃N₃O₂ MW: 323.7 g/mol Melting Pt: 75…76 °C |
MDL Number:
MFCD12025894 CAS Number: 1147979-45-0 |
Specification Test Results
Melting point | 75-76°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...