2-(4-Methoxy-3-nitrophenyl)acetohydrazide ≥95%

Supplier: Matrix Scientific

046669-500MG 046669-1G 046669-5G
101876-484EA 375.38 USD
101876-484 101876-486 101876-488
2-(4-Methoxy-3-nitrophenyl)acetohydrazide ≥95%
2-(4-Methoxy-3-nitrophenyl)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁N₃O₄
Melting Pt: 158…159 °C
MDL Number: MFCD09754969
CAS Number: 944892-06-2

Order Now

Specification Test Results

Melting point 158-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR