(2,3,5-Trichloro-4-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

046668-500MG 046668-1G
101876-480EA 921.54 USD
101876-480 101876-482
(2,3,5-Trichloro-4-pyridinyl)methanol ≥95%
(2,3,5-Trichloro-4-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₄Cl₃NO
Melting Pt: 74…75 °C
MDL Number: MFCD12025893
CAS Number: 1147979-44-9

Order Now

Specification Test Results

Melting point 74-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR