Methyl-2-(acetylamino)-5-bromo-3-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
046667-500MG
046667-1G
046667-5G
101876-474EA
423.08
USD
101876-474
101876-476
101876-478
Methyl-2-(acetylamino)-5-bromo-3-thiophenecarboxylate ≥95%
Methyl-2-(acetylamino)-5-bromo-3-thiophenecarboxylate
Hazard: Irritant
Formula:
C₈H₈BrNO₃S Melting Pt: 112…114 °C |
MDL Number:
MFCD06408953 CAS Number: 874816-04-3 |
Specification Test Results
Melting point | 112-114°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...