3,5-Dichloro-2-methoxyisonicotinic acid ≥95%

Supplier: Matrix Scientific

046659-500MG 046659-1G
101876-446EA 747.69 USD
101876-446 101876-448
3,5-Dichloro-2-methoxyisonicotinic acid ≥95%
3,5-Dichloro-2-methoxyisonicotinic acid

Hazard: Irritant

Formula: C₇H₅Cl₂NO₃
MW: 222.025 g/mol
Melting Pt: 151-152°C
MDL Number: MFCD12025889
CAS Number: 1147979-40-5

Order Now

Specification Test Results

Melting point 151-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR