4-Chloro-5-methoxy-2-(3-nitrophenyl)pyrimidine ≥95%

Supplier: Matrix Scientific

046656-500MG 046656-1G
101876-436EA 1109.23 USD
101876-436 101876-438
4-Chloro-5-methoxy-2-(3-nitrophenyl)pyrimidine ≥95%
4-Chloro-5-methoxy-2-(3-nitrophenyl)pyrimidine

Hazard: Irritant

Formula: C₁₁H₈ClN₃O₃
Melting Pt: 131…133 °C
MDL Number: MFCD12025887
CAS Number: 1147979-38-1

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR