5-Methoxy-2-(3-nitrophenyl)-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
046654-500MG
046654-1G
101876-428EA
921.54
USD
101876-428
101876-430
5-Methoxy-2-(3-nitrophenyl)-4-pyrimidinol ≥95%
5-Methoxy-2-(3-nitrophenyl)-4-pyrimidinol
Hazard: Irritant
Formula:
C₁₁H₉N₃O₄ Melting Pt: 280 °C (dec.) |
MDL Number:
MFCD12025886 CAS Number: 1147979-37-0 |
Specification Test Results
Melting point | 280°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...