Methyl thieno[2,3-b]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
046653-500MG
046653-1G
046653-5G
101876-422EA
375.38
USD
101876-422
101876-424
101876-426
Methyl thieno[2,3-b]pyridine-2-carboxylate ≥95%
Methyl thieno[2,3-b]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₉H₇NO₂S Melting Pt: 106…107 °C |
MDL Number:
MFCD12025885 CAS Number: 154650-88-1 |
Specification Test Results
Melting point | 106-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...