Methyl-3-chloro-2-methoxy-5-(trifluoromethyl)isonicotinate ≥95%
Supplier: Matrix Scientific
046652-500MG
046652-1G
101876-418EA
921.54
USD
101876-418
101876-420
Methyl-3-chloro-2-methoxy-5-(trifluoromethyl)isonicotinate ≥95%
Methyl-3-chloro-2-methoxy-5-(trifluoromethyl)isonicotinate
Hazard: Irritant
Formula:
C₉H₇ClF₃NO₃ Melting Pt: 49…50 °C |
MDL Number:
MFCD12025884 CAS Number: 1147979-36-9 |
Specification Test Results
Melting point | 49-50°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...