2-Bromo-6-(trifluoromethyl)nicotinic acid ≥95%

Supplier: Matrix Scientific

046648-500MG 046648-1G
101876-402EA 747.69 USD
101876-402 101876-404
2-Bromo-6-(trifluoromethyl)nicotinic acid ≥95%
2-Bromo-6-(trifluoromethyl)nicotinic acid

Hazard: Irritant

Formula: C₇H₃BrF₃NO₂
Melting Pt: 130…132 °C
MDL Number: MFCD09909645
CAS Number: 749875-07-8

Order Now

Specification Test Results

Melting point 130-132°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR