Ethyl-5-bromo-3-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

046646-500MG 046646-1G
101876-394EA 747.69 USD
101876-394 101876-396
Ethyl-5-bromo-3-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylate ≥95%
Ethyl-5-bromo-3-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₈BrNO₅
Melting Pt: 128…130 °C
MDL Number: MFCD12025880
CAS Number: 1160474-41-8

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR