2-(3,4-Dichlorophenyl)-3-hydroxyacrylonitrile ≥95%

Supplier: Matrix Scientific

046643-500MG 046643-1G
101876-382EA 278.46 USD
101876-382 101876-384
2-(3,4-Dichlorophenyl)-3-hydroxyacrylonitrile ≥95%
2-(3,4-Dichlorophenyl)-3-hydroxyacrylonitrile

Hazard: Irritant

Formula: C₉H₅Cl₂NO
Melting Pt: 164…166 °C
MDL Number: MFCD12025879
CAS Number: 77186-41-5

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR