3-Hydroxy-2-(2-thienyl)acrylonitrile ≥95%
Supplier: Matrix Scientific
046641-500MG
046641-1G
101876-374EA
278.46
USD
101876-374
101876-376
3-Hydroxy-2-(2-thienyl)acrylonitrile ≥95%
3-Hydroxy-2-(2-thienyl)acrylonitrile
Hazard: Irritant
Formula:
C₇H₅NOS Melting Pt: 136…137 °C |
MDL Number:
MFCD01859930 CAS Number: 71637-32-6 |
Specification Test Results
Melting point | 136-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...