3-Hydroxy-2-[3-(trifluoromethyl)phenyl]acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-[3-(trifluoromethyl)phenyl]prop-2-enenitrile

046640-500MG 046640-1G
101876-370EA 278.46 USD
101876-370 101876-372
3-Hydroxy-2-[3-(trifluoromethyl)phenyl]acrylonitrile ≥95%
3-Hydroxy-2-[3-(trifluoromethyl)phenyl]acrylonitrile

Hazard: Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 96…97 °C
MDL Number: MFCD12025877
CAS Number: 62739-04-2

Order Now

Specification Test Results

Melting point 96-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR