Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,5,6-trichloropyrimidine-4-carboxylate

046639-500MG 046639-1G
101876-366EA 738.46 USD
101876-366 101876-368
Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate ≥95%
Methyl-2,5,6-trichloro-4-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₆H₃Cl₃N₂O₂
MW: 241.46 g/mol
Melting Pt: 46…49 °C
MDL Number: MFCD12025876
CAS Number: 89284-85-5

Order Now

Specification Test Results

Melting point 46-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR