2-Amino-5-iodo-6-methyl-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046638-500MG 046638-1G
101876-362EA 338.46 USD
101876-362 101876-364
2-Amino-5-iodo-6-methyl-4-pyrimidinol ≥95%
2-Amino-5-iodo-6-methyl-4-pyrimidinol

Hazard: Irritant

Formula: C₅H₆IN₃O
Melting Pt: 219…220 °C
MDL Number: MFCD12025875
CAS Number: 22294-57-1

Order Now

Specification Test Results

Melting point 219-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR