4-Chloro-5-iodo-6-methyl-2-pyrimidinamine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-4-chloro-5-iodo-6-methylpyrimidine
046637-500MG
046637-1G
101876-358EA
295.38
USD
101876-358
101876-360
4-Chloro-5-iodo-6-methyl-2-pyrimidinamine ≥95%
4-Chloro-5-iodo-6-methyl-2-pyrimidinamine
Hazard: Irritant
Formula:
C₅H₅ClIN₃ MW: 269.47 g/mol Melting Pt: 197…199 °C |
MDL Number:
MFCD12024467 CAS Number: 897030-99-8 |
Specification Test Results
Melting point | 197-199°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...