2-Chloro-6-methylisonicotinic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-6-methylpyridine-4-carboxylic acid
041238-5G
041238-25G
101851-012EA
467.69
USD
101851-012
102833-110
2-Chloro-6-methylisonicotinic acid ≥97%
2-Chloro-6-methylisonicotinic acid
Hazard: Irritant
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD00052830 CAS Number: 25462-85-5 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...