2-Amino-N,N-dimethylbenzamide

Supplier: Matrix Scientific

023912-1G 023912-5G
101817-022EA 32.31 USD
101817-022 101817-024
2-Amino-N,N-dimethylbenzamide
2-Amino-N,N-dimethylbenzamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
Melting Pt: 54…56 °C
MDL Number: MFCD01168906
CAS Number: 6526-66-5

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR