Methyl-3,5-dihydroxyphenylacetate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-(3,5-dihydroxyphenyl)acetate
023795-1G
023795-5G
101816-764EA
116.92
USD
101816-764
101816-766
Methyl-3,5-dihydroxyphenylacetate ≥97%
Methyl-3,5-dihydroxyphenylacetate
Hazard: Irritant
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol Melting Pt: 109…112 °C |
MDL Number:
MFCD00091620 CAS Number: 4724-10-1 |
Specification Test Results
Melting point | 109-112°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...