5-Bromo-2-methoxynicotinic acid ≥99%

Supplier: Matrix Scientific

023784-1G 023784-5G
101816-736EA 21.54 USD
101816-736 101816-738
5-Bromo-2-methoxynicotinic acid ≥99%
5-Bromo-2-methoxynicotinic acid

Hazard: Irritant

Formula: C₇H₆BrNO₃
Melting Pt: 158-158.5°C
MDL Number: MFCD06656771
CAS Number: 54916-66-4

Order Now

Specification Test Results

Melting point 158-158.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR