Methyl-4-chloropicolinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 4-chloropicolinate, Methyl 4-chloropyridine-2-carboxylate

023779-5G 023779-25G
101816-724EA 38.46 USD
101816-724 101816-726
Methyl-4-chloropicolinate ≥97%
Methyl 4-chloropicolinate

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
Melting Pt: 58-59°C
MDL Number: MFCD04116183
CAS Number: 24484-93-3

Order Now

Specification Test Results

Melting point 58-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR