5-Bromo-7-methyl-1H-indazole ≥97%
Supplier: Matrix Scientific
023773-250MG
023773-1G
101816-702EA
67.69
USD
101816-702
101816-704
5-Bromo-7-methyl-1H-indazole ≥97%
5-Bromo-7-methyl-1H-indazole
Hazard: Irritant
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol Melting Pt: 184…185 °C |
MDL Number:
MFCD03990484 CAS Number: 156454-43-2 |
Specification Test Results
Melting point | 184-185°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...