3-Chloro-4-fluorobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

023770-1G 023770-5G
101816-690EA 36.92 USD
101816-690 101816-692
3-Chloro-4-fluorobenzenesulfonamide ≥97%
3-Chloro-4-fluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₅ClFNO₂S
Melting Pt: 108-109°C
MDL Number: MFCD00130211
CAS Number: 146533-46-2

Order Now

Specification Test Results

Melting point 108-109°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR