(4-(Trifluoromethyl)phenyl)methanesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: [4-(Trifluoromethyl)phenyl]methanesulphonyl chloride, 4-Trifluoromethyl-α-toluenesulphonyl chloride

023764-1G 023764-5G
101816-672EA 53.85 USD
101816-672 101816-674
(4-(Trifluoromethyl)phenyl)methanesulfonyl chloride ≥97%
(4-(Trifluoromethyl)phenyl)methanesulphonyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₈H₆ClF₃O₂S
MW: 258.65 g/mol
Melting Pt: 103-107°C
MDL Number: MFCD01631932
CAS Number: 163295-75-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 105-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR