5-(Trifluoromethyl)picolinic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-(Trifluoromethyl)pyridine-2-carboxylic acid
023761-1G
023761-5G
101816-664EA
15.38
USD
101816-664
101816-666
5-(Trifluoromethyl)picolinic acid ≥95%
5-(Trifluoromethyl)picolinic acid
Hazard: Irritant
Formula:
C₇H₄F₃NO₂ MW: 191.11 g/mol Melting Pt: 135-137°C |
MDL Number:
MFCD04113632 CAS Number: 80194-69-0 |
Specification Test Results
Melting point | 135-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...