6-Chloro-3-pyridineacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(6-Chloropyridin-3-yl)acetic acid

023734-5G 023734-25G
101816-594EA 44.62 USD
101816-594 101816-596
6-Chloro-3-pyridineacetic acid ≥98%
6-Chloro-3-pyridineacetic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 164…169 °C
MDL Number: MFCD01863172
CAS Number: 39891-13-9

Order Now

Specification Test Results

Melting point 164-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR