(4-Chloro-2-pyridinyl)methanol ≥98%
Supplier: Matrix Scientific
Synonyms:
(4-Chloropyridin-2-yl)methanol, 4-Chloro-2-(hydroxymethyl)pyridine
023726-1G
023726-5G
101816-578EA
15.38
USD
101816-578
101816-580
(4-Chloro-2-pyridinyl)methanol ≥98%
(4-Chloro-2-pyridinyl)methanol
Hazard: Irritant
Formula:
C₆H₆ClNO MW: 143.5709 g/mol Melting Pt: 49-51°C |
MDL Number:
MFCD07437885 CAS Number: 63071-10-3 |
Specification Test Results
Melting point | 49-51°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...