D-Phenylalaninol ≥95%

Supplier: Matrix Scientific

Synonyms: D-Penylalaninol, D(+)-2-Amino-3-phenyl-1-propanol

021578-25G 021578-100G
101812-348EA 64.62 USD
101812-348 101812-350
D-Phenylalaninol ≥95%
D-Phenylalaninol

Hazard: Irritant

Formula: C₉H₁₃NO
MW: 151.2 g/mol
Melting Pt: 92…93 °C
MDL Number: MFCD00064298
CAS Number: 5267-64-1

Order Now

Specification Test Results

Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR