3-Amino-α,α,α-trifluoro-p-toluic acid ≥98%

Supplier: Matrix Scientific

021433-1G 021433-5G 021433-25G
101812-094EA 41.54 USD
101812-094 101812-096 101812-098
3-Amino-α,α,α-trifluoro-p-toluic acid ≥98%
3-Amino-α,α,α-trifluoro-p-toluic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₂
Melting Pt: 203…205 °C
MDL Number: MFCD07784307
CAS Number: 125483-00-3

Order Now

Specification Test Results

Melting point 203-205°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR