(3-Methoxyphenyl)hydrazine hydrochloride ≥98%
Supplier: Matrix Scientific
021395-25G
021395-100G
101811-960EA
306.15
USD
101811-960
101811-962
(3-Methoxyphenyl)hydrazine hydrochloride ≥98%
(3-Methoxyphenyl)hydrazine hydrochloride
Hazard: Irritant/Store in Freezer
Formula:
C₇H₁₁ClN₂O Melting Pt: 142 °C (dec.) |
MDL Number:
MFCD00044416 CAS Number: 39232-91-2 |
Specification Test Results
Melting point | 142°C(dec) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...