5-Chloro-2-fluorobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

021394-1G 021394-5G
101811-954EA 38.46 USD
101811-954 101811-956
5-Chloro-2-fluorobenzenesulfonamide ≥97%
5-Chloro-2-fluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₅ClFNO₂S
MW: 209.63 g/mol
Melting Pt: 134-136°C
MDL Number: MFCD03094289
CAS Number: 351003-57-1

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR