4-Bromonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromopyridine-3-carboxylic acid

021379-1G 021379-5G
101811-910EA 38.46 USD
101811-910 101811-912
4-Bromonicotinic acid ≥97%
4-Bromonicotinic acid

Hazard: Irritant/Stored under Argon/Store in Freezer

Formula: C₆H₄BrNO₂
MW: 202.007 g/mol
Melting Pt: 165…166 °C (dec.)
MDL Number: MFCD04114236
CAS Number: 15366-62-8

Order Now

Specification Test Results

Melting point 165-166°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR