(4-Methylphenyl)methanesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
p-Tolylmethanesulphonyl chloride, α-p-Xylenesulphonyl chloride, 4-Methylbenzylsulphonyl chloride
021361-1G
021361-5G
101811-844EA
73.85
USD
101811-844
101811-846
(4-Methylphenyl)methanesulfonyl chloride ≥97%
(4-Methylphenyl)methanesulphonyl chloride
Hazard: Irritant/Store in Freezer
Formula:
C₈H₉ClO₂S MW: 204.67 g/mol Melting Pt: 80°C |
MDL Number:
MFCD01631931 CAS Number: 51419-59-1 |
Specification Test Results
Melting point | 80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...