7-Azaindole-5-boronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

Synonyms: 7-Azaindole-5-boronic Acid Pinacol Ester

021144-250MG 021144-1G
101811-442EA 52.31 USD
101811-442 101811-444
7-Azaindole-5-boronic acid pinacol ester ≥98%
7-Azaindole-5-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₃H₁₇BN₂O₂
MW: 244.101 g/mol
Melting Pt: 242…242.5 °C
MDL Number: MFCD08060937
CAS Number: 754214-56-7

Order Now

Specification Test Results

Melting point 242-242.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR