2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine ≥99%

Supplier: Matrix Scientific

Synonyms: 2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide)

021135-1G 021135-5G
101811-416EA 38.46 USD
101811-416 101811-418
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine ≥99%
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine

Hazard: Irritant-Harmful/Store in Freezer/Air Sensitive

Formula: C₇H₃ClF₆N₂O₄S₂
MW: 392.67 g/mol
Melting Pt: 45…47 °C
MDL Number: MFCD00191833
CAS Number: 145100-51-2

Order Now

Specification Test Results

Melting point 45-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR