5-Chloro-2-methoxy-4-methylbenzenesulfonamide ≥97%

Supplier: Matrix Scientific

021122-1G 021122-5G
101811-376EA 63.08 USD
101811-376 101811-378
5-Chloro-2-methoxy-4-methylbenzenesulfonamide ≥97%
5-Chloro-2-methoxy-4-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₈H₁₀ClNO₃S
Melting Pt: 173…175 °C
MDL Number: MFCD08445708
CAS Number: 199590-75-5

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR